KEY004181677 Key Organics / BIONET

2-(prop-2-yn-1-yloxy)benzaldehyde

  • MDL number MFCD02380378
  • Cas Number : 29978-83-4
  • Molecular Weight : 160.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 68
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93574

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0262_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart