KEY004181678 Key Organics / BIONET

2-hydroxybenzene-1,3,5-tricarbaldehyde

  • MDL number MFCD01314207
  • Cas Number : 81502-74-1
  • Molecular Weight : 178.143

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201 - 202
Purity 95
Storage temp. Store under N2
Physical Form Off-white to Dark Brown Solid
Shipping Temperature Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93585

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0275_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart