KEY004181679 Key Organics / BIONET

[2-(prop-2-yn-1-yloxy)phenyl]methanol

  • MDL number MFCD03001302
  • Cas Number : 130441-68-8
  • Molecular Weight : 162.188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38 - 40
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93589

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart