KEY004181682 Key Organics / BIONET

1-(4-acetyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

  • MDL number MFCD03001304
  • Cas Number : 72652-34-7
  • Molecular Weight : 254.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 201 - 203
Iupac Name 1-(4-acetyl-1H-pyrrol-2-yl)-2,2,2-trichloroethanone
Inchi Code 1S/C8H6Cl3NO2/c1-4(13)5-2-6(12-3-5)7(14)8(9,10)11/h2-3,12H,1H3
Inchi Key OEXMOGNTZJXVPZ-UHFFFAOYSA-N

Description

Melting Point 201 - 203
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93596

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0288_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart