KEY004181684 Key Organics / BIONET

5-chloro-2-(prop-2-yn-1-yloxy)benzaldehyde

  • MDL number MFCD03001306
  • Cas Number : 224317-64-0
  • Molecular Weight : 194.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 76
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93610

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0306_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart