KEY004181688 Key Organics / BIONET

3-(2,4-dichlorophenyl)-1-oxo-3,4-dihydro-1H-2-benzopyran-4-carboxylic acid

  • MDL number MFCD03001309
  • Cas Number : 400751-70-4
  • Molecular Weight : 337.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216 - 219
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93674

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0381_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart