KEY004181689 Key Organics / BIONET

3-(thiophen-2-yl)-3-(2,2,2-trifluoroacetamido)propanoic acid

  • MDL number MFCD03001310
  • Cas Number : 115957-22-7
  • Molecular Weight : 267.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 157
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93684

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0606_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart