KEY004181692 Key Organics / BIONET

3-(2H-1,3-benzodioxol-5-yl)-3-{[(tert-butoxy)carbonyl]amino}propanoic acid

  • MDL number MFCD02090675
  • Cas Number : 149520-06-9
  • Molecular Weight : 309.318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 160
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93689

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0611_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart