KEY004181698 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(3-hydroxyphenyl)propanoic acid

  • MDL number MFCD02090711
  • Cas Number : 284493-68-1
  • Molecular Weight : 281.308

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93715

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0641_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart