KEY004181699 Key Organics / BIONET

3-{[(benzyloxy)carbonyl]amino}-3-(thiophen-2-yl)propanoic acid

  • MDL number MFCD03001313
  • Cas Number : 73457-03-1
  • Molecular Weight : 305.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93725

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0656_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart