KEY007384642 Key Organics / BIONET

1-methyl-4-(4-nitrophenyl)piperazine

  • MDL number MFCD00101003
  • Cas Number : 16155-03-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-419S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart