KEY007384643 Key Organics / BIONET

3-{[(4-nitrophenyl)methyl]sulfanyl}thiophene-2-carboxylic acid

  • MDL number MFCD01316316
  • Cas Number : 251097-01-5
  • Molecular Weight : 295.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198 - 201
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/797

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-439S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart