KEY007384652 Key Organics / BIONET

1-methyl-4-(4-nitrobenzoyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD00172169
  • Cas Number : 128843-59-4
  • Molecular Weight : 258.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 212
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/938

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10H-022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart