KEY007384683 Key Organics / BIONET

3-(2-fluoro-4-nitrophenoxy)thiophene-2-carboxylic acid

  • MDL number MFCD00202749
  • Cas Number : 337919-93-4
  • Molecular Weight : 283.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135 - 137
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1155

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10H-602S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart