KEY007384862 Key Organics / BIONET

3-(4-fluoro-2-nitrophenoxy)thiophene-2-carboxylic acid

  • MDL number MFCD00214992
  • Cas Number : 303152-17-2
  • Molecular Weight : 283.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159 - 161
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4700

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11H-325S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart