KEY007385036 Key Organics / BIONET

3-(4-nitrophenyl)-3-(2,2,2-trifluoroacetamido)propanoic acid

  • MDL number MFCD02661624
  • Cas Number : 439094-19-6
  • Molecular Weight : 306.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 170
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6744

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0627_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart