KEY007385071 Key Organics / BIONET

ethyl 1-(4-cyanophenyl)piperidine-4-carboxylate

  • MDL number MFCD03086198
  • Cas Number : 352018-90-7
  • Molecular Weight : 258.321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71 - 72
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6884

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0236_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart