KEY007385116 Key Organics / BIONET

5-bromo-7-nitro-1H-indazole

  • MDL number MFCD03617604
  • Cas Number : 316810-82-9
  • Molecular Weight : 242.032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7043

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0823_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart