KEY007385203 Key Organics / BIONET

5-methyl-1-(4-nitrophenyl)-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD00173063
  • Cas Number : 1025-88-3
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 203 - 205
Iupac Name 5-methyl-1-(4-nitrophenyl)-1H-1,2,4-triazole-3-carboxylic acid
Inchi Code 1S/C10H8N4O4/c1-6-11-9(10(15)16)12-13(6)7-2-4-8(5-3-7)14(17)18/h2-5H,1H3,(H,15,16)
Inchi Key AGPQLGNAAHYBPG-UHFFFAOYSA-N

Description

Melting Point 203 - 205
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8053

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12G-348S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart