KEY007385298 Key Organics / BIONET

1,3-dimethyl 2-(2-nitrophenyl)propanedioate

  • MDL number MFCD03617780
  • Cas Number : 26465-37-2
  • Molecular Weight : 253.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48 - 50
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9734

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart