KEY007385370 Key Organics / BIONET

4-(4-chlorophenoxy)-3-nitrobenzaldehyde

  • MDL number MFCD01113126
  • Cas Number : 320416-35-1
  • Molecular Weight : 277.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63300

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1D-047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart