KEY007385372 Key Organics / BIONET

methyl 4-(4-chlorophenoxy)-3-nitrobenzoate

  • MDL number MFCD00138745
  • Cas Number : 320416-58-8
  • Molecular Weight : 307.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125 - 127
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63318

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1D-067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart