KEY007385382 Key Organics / BIONET

4-(3-chlorophenoxy)-3-nitrobenzaldehyde

  • MDL number MFCD00138762
  • Cas Number : 320417-03-6
  • Molecular Weight : 277.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 80 - 82
Iupac Name 4-(3-chlorophenoxy)-3-nitrobenzaldehyde
Inchi Code 1S/C13H8ClNO4/c14-10-2-1-3-11(7-10)19-13-5-4-9(8-16)6-12(13)15(17)18/h1-8H
Inchi Key FLWAMJZFOINMFU-UHFFFAOYSA-N

Description

Melting Point 80 - 82
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63343

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1D-113_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart