KEY007385552 Key Organics / BIONET

(2E)-1-(5-bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD00098108
  • Cas Number : 938176-46-6
  • Molecular Weight : 260.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 118
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65851

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0633_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart