KEY007385553 Key Organics / BIONET

(2E)-1-(5-chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD03617978
  • Cas Number : 1276536-34-5
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65852

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0634_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart