KEY007385554 Key Organics / BIONET

4-(5-chlorothiophen-2-yl)pyrimidin-2-amine

  • MDL number MFCD03617979
  • Cas Number : 855308-65-5
  • Molecular Weight : 211.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65861

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0646_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart