KEY007385606 Key Organics / BIONET

4-(pyrimidin-2-yloxy)benzoic acid

  • MDL number MFCD03617910
  • Cas Number : 855423-33-5
  • Molecular Weight : 216.196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0313_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart