KEY007385655 Key Organics / BIONET

ethyl 5-oxo-2H,3H,5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylate

  • MDL number MFCD00085067
  • Cas Number : 32084-53-0
  • Molecular Weight : 226.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175 - 177
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66166

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0834_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart