KEY007385876 Key Organics / BIONET

2-(2-hydroxyethyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one

  • MDL number MFCD03618033
  • Cas Number : 191097-21-9
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 149
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0332_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart