KEY007386036 Key Organics / BIONET

2-amino-6-(4-nitrophenyl)-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carbonitrile

  • MDL number MFCD00794642
  • Cas Number : 338413-84-6
  • Molecular Weight : 300.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240 - 242
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71792

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-910_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart