KEY007386060 Key Organics / BIONET

2-(2,4-dinitrophenyl)cyclohexan-1-one

  • MDL number MFCD02186041
  • Cas Number : 21442-55-7
  • Molecular Weight : 264.237

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 101.5
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72567

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0086_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart