KEY007386230 Key Organics / BIONET

2-[4-(4-nitrophenyl)piperazin-1-yl]ethan-1-ol

  • MDL number MFCD00511837
  • Cas Number : 5521-38-0
  • Molecular Weight : 251.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 97 - 99
Iupac Name 2-[4-(4-nitrophenyl)-1-piperazinyl]ethanol
Inchi Code 1S/C12H17N3O3/c16-10-9-13-5-7-14(8-6-13)11-1-3-12(4-2-11)15(17)18/h1-4,16H,5-10H2
Inchi Key WVNPWSRYQDRSIS-UHFFFAOYSA-N

Description

Melting Point 97 - 99
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75754

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-901_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart