KEY007386434 Key Organics / BIONET

4-(2,4-difluorophenoxy)-3-nitrobenzaldehyde

  • MDL number MFCD00794806
  • Cas Number : 173282-22-9
  • Molecular Weight : 279.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 88 - 90
Iupac Name 4-(2,4-difluorophenoxy)-3-nitrobenzaldehyde
Inchi Code 1S/C13H7F2NO4/c14-9-2-4-12(10(15)6-9)20-13-3-1-8(7-17)5-11(13)16(18)19/h1-7H
Inchi Key RINTXUSUROIDBP-UHFFFAOYSA-N

Description

Melting Point 88 - 90
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79218

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5L-501S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart