KEY007386585 Key Organics / BIONET

(Z)-N'-hydroxy-1-(1,3-thiazol-2-yl)-1H-pyrrole-2-carboximidamide

  • MDL number MFCD03617498
  • Cas Number : 685107-91-9
  • Molecular Weight : 208.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 170DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80649

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0639_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart