KEY007386685 Key Organics / BIONET

6-(4-nitrophenyl)-5H,6H,7H,8H-pyrido[4,3-d]pyrimidin-2-amine

  • MDL number MFCD00180838
  • Cas Number : 338791-45-0
  • Molecular Weight : 271.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262 - 265
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81673

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6F-925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart