KEY007386760 Key Organics / BIONET

2-[(2-chloro-1,3-thiazol-5-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD03617669
  • Cas Number : 120740-09-2
  • Molecular Weight : 278.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107 - 108
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83935

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart