KEY007386846 Key Organics / BIONET

1-(4-nitrophenyl)pyrrolidin-2-one

  • MDL number MFCD00138492
  • Cas Number : 13691-26-4
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 129 - 130
Iupac Name 1-(4-nitrophenyl)-2-pyrrolidinone
Inchi Code 1S/C10H10N2O3/c13-10-2-1-7-11(10)8-3-5-9(6-4-8)12(14)15/h3-6H,1-2,7H2
Inchi Key YYYMDBUHBOEDTC-UHFFFAOYSA-N

Description

Melting Point 129 - 130
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84642

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7C-147_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart