KEY007387128 Key Organics / BIONET

methyl 4-{[2-nitro-4-(trifluoromethyl)phenyl]amino}butanoate

  • MDL number MFCD00793100
  • Cas Number : 339101-25-6
  • Molecular Weight : 306.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81 - 83
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89713

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8L-915_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart