KEY007387129 Key Organics / BIONET

4-[(3-nitropyridin-2-yl)amino]butanoic acid

  • MDL number MFCD00793101
  • Cas Number : 339101-31-4
  • Molecular Weight : 225.204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89715

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8L-917_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart