KEY007387171 Key Organics / BIONET

(5Z)-5-[(3-nitrophenyl)methylidene]-1,3-thiazolidine-2,4-dione

  • MDL number MFCD00817881
  • Cas Number : 24044-52-8
  • Molecular Weight : 250.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 187
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90883

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0073_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart