KEY007387349 Key Organics / BIONET

2-[(2E)-3-(4-nitrophenyl)prop-2-enoyl]benzoic acid

  • MDL number MFCD03617865
  • Cas Number : 6261-66-1
  • Molecular Weight : 297.266

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187 - 189
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93902

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9T-0056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart