KEY007387369 Key Organics / BIONET

1-[(4-chlorophenyl)methyl]-1H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD01175876
  • Cas Number : 31694-94-7
  • Molecular Weight : 209.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201 - 203
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10A-024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart