KEY007387372 Key Organics / BIONET

2-amino-1-benzyl-4,5-dimethyl-1H-pyrrole-3-carbonitrile

  • MDL number MFCD01935932
  • Cas Number : 55817-72-6
  • Molecular Weight : 225.295

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152 - 155
Purity 95
Storage temp. Store under N2
Physical Form Off-white to Black Solid
Shipping Temperature Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1706

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/10L-318S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart