KEY007387378 Key Organics / BIONET

ethyl 4-(2,5-dimethoxyphenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD00397696
  • Cas Number : 292855-70-0
  • Molecular Weight : 320.345

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218 - 219
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0327_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart