KEY007387386 Key Organics / BIONET

ethyl 4-(4-methoxyphenyl)-2-oxo-6-phenylcyclohex-3-ene-1-carboxylate

  • MDL number MFCD02031258
  • Cas Number : 25960-25-2
  • Molecular Weight : 350.414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107 - 108
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3093

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0218_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart