KEY007387387 Key Organics / BIONET

5-(4-pentylcyclohexyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD03791141
  • Cas Number : 446275-94-1
  • Molecular Weight : 254.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3096

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0222_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart