KEY007387389 Key Organics / BIONET

ethyl (2E)-3-[4-(2,4-dichlorobenzoyl)-1-methyl-1H-pyrrol-2-yl]prop-2-enoate

  • MDL number MFCD02571785
  • Cas Number : 400077-69-2
  • Molecular Weight : 352.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 89
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3114

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0250_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart