KEY007387390 Key Organics / BIONET

ethyl 4-(2-methoxyphenyl)-2-oxo-6-(thiophen-3-yl)cyclohex-3-ene-1-carboxylate

  • MDL number MFCD04113617
  • Cas Number : 446275-96-3
  • Molecular Weight : 356.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145 - 147
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3116

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0254_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart