KEY007387392 Key Organics / BIONET

4-hydroxy-3-propyl-1-azatricyclo[7.3.1.0^{5,13}]trideca-3,5(13),6,8-tetraen-2-one

  • MDL number MFCD03791142
  • Cas Number : 57625-51-1
  • Molecular Weight : 243.306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 172
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3128

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0272_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart