KEY007387394 Key Organics / BIONET

ethyl 4,6-bis(4-chlorophenyl)-2-oxocyclohex-3-ene-1-carboxylate

  • MDL number MFCD03425713
  • Cas Number : 26379-96-4
  • Molecular Weight : 389.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3145

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0298_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart